Gene Transcripts

Gene Symbol
Cisd2
Entrez Gene ID
295457
Gene Name
CDGSH iron sulfur domain 2
Gene Aliases
RGD1566242
Chromosome Location
Chr. 2: 240586754 - 240611560 on Build Rnor_6.0
UniGene ID
Rn.24858
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001191608.1 NP_001178537.1 2 308

View Full Product Details