Gene Transcripts

Gene Symbol
Phospho2
Entrez Gene ID
295663
Gene Name
phosphatase, orphan 2
Gene Aliases
Chromosome Location
Chr. 3: 56030915 - 56038076 on Build Rnor_6.0
UniGene ID
Rn.1266
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001007642.1 NP_001007643.1 4 614
GenBank mRNA BC081926.1 - 4 614

View Full Product Details