Gene Transcripts

Gene Symbol
Zc3hc1
Entrez Gene ID
296957
Gene Name
zinc finger, C3HC-type containing 1
Gene Aliases
RGD1561099
Chromosome Location
Chr. 4: 57684678 - 57706613 on Build Rnor_6.0
UniGene ID
Rn.56620
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001127293.1 NP_001120765.1 6 721
GenBank mRNA BC166834.1 - 6 721

View Full Product Details