Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001106612.1 | NP_001100082.1 | 4 | 331 | |
XM_008763159.1 | XP_008761381.1 | 4 | 745 | ||
XM_008763160.1 | XP_008761382.1 | 6 | 815 | ||
XM_008763161.1 | XP_008761383.1 | 5 | 679 | ||
GenBank mRNA | BC168936.1 | - | 4 | 318 |