Gene Transcripts

Gene Symbol
Shq1
Entrez Gene ID
297483
Gene Name
SHQ1, H/ACA ribonucleoprotein assembly factor
Gene Aliases
RGD1310610
Chromosome Location
Chr. 4: 133037515 - 133127183 on Build Rnor_6.0
UniGene ID
Rn.54034
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001134713.1 NP_001128185.1 602
XM_008763162.1 XP_008761384.1 610
GenBank mRNA BC168172.1 - 602

View Full Product Details