Gene Transcripts

Gene Symbol
Mysm1
Entrez Gene ID
298247
Gene Name
myb-like, SWIRM and MPN domains 1
Gene Aliases
RGD1311787
Chromosome Location
Chr. 5: 113906409 - 113939085 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_006238409.2 XP_006238471.1 5, 6 752
XM_008763911.1 XP_008762133.1 5, 6 371
XM_008763912.1 XP_008762134.1 5, 6 371
XM_008763913.1 XP_008762135.1 5, 6 371

View Full Product Details