Gene Transcripts

Gene Symbol
Aim1l
Entrez Gene ID
298543
Gene Name
absent in melanoma 1-like
Gene Aliases
Chromosome Location
Chr. 5: 152289490 - 152321539 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_008764189.1 XP_008762411.1 3 1294

View Full Product Details