Gene Transcripts

Gene Symbol
Isg15
Entrez Gene ID
298693
Gene Name
ISG15 ubiquitin-like modifier
Gene Aliases
G1p2
Chromosome Location
Chr. 5: 173624862 - 173629131 on Build Rnor_6.0
UniGene ID
Rn.198318
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001106700.1 NP_001100170.1 2 391
XM_006239563.2 XP_006239625.1 3 985

View Full Product Details