Gene Transcripts
- Gene Symbol
- Acot6
- Entrez Gene ID
- 299193
- Gene Name
- acyl-CoA thioesterase 6
- Gene Aliases
- RGD1309669
- Chromosome Location
- Chr. 6: 107581285 - 107590471 on Build Rnor_6.0
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
XM_234402.7 |
XP_234402.5
|
3 |
1306 |
|
View Full Product Details