Gene Transcripts

Gene Symbol
Dnal4
Entrez Gene ID
300078
Gene Name
dynein, axonemal, light chain 4
Gene Aliases
Dnalc4
Chromosome Location
Chr. 7: 120987438 - 121000272 on Build Rnor_6.0
UniGene ID
Rn.3257
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001009666.1 NP_001009666.1 4 399
GenBank mRNA BC087688.1 - 4 399

View Full Product Details