Gene Transcripts

Gene Symbol
Dusp7
Entrez Gene ID
300980
Gene Name
dual specificity phosphatase 7
Gene Aliases
mkp-X
Chromosome Location
Chr. 8: 115069095 - 115076130 on Build Rnor_6.0
UniGene ID
Rn.104502
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001100547.1 NP_001094017.1 2 845
GenBank mRNA X94186.1 - 2 428

View Full Product Details