Gene Transcripts

Gene Symbol
AQP9
Entrez Gene ID
366
Gene Name
aquaporin 9
Gene Aliases
AQP-9, HsT17287, SSC1, T17287
Chromosome Location
Chr. 15: 58138169 - 58185911 on Build GRCh38
UniGene ID
Hs.104624
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001320635.1 NP_001307564.1 5 974
NM_001320636.1 NP_001307565.1 6 824
NM_020980.4 NP_066190.2 6 1192
GenBank mRNA AB008775.1 - 6 1038
AF016495.1 - 6 1050
AK300055.1 - 6 797
AK313214.1 - 6 1117
BC026258.1 - 6 1007 4734559
CR542027.1 - 6 822

View Full Product Details