Gene Transcripts

Gene Symbol
IFRD2
Entrez Gene ID
7866
Gene Name
interferon related developmental regulator 2
Gene Aliases
IFNRP, SKMc15, SM15
Chromosome Location
Chr. 3: 50287732 - 50292595 on Build GRCh38
UniGene ID
Hs.315177
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_006764.4 NP_006755.4 7 1070
GenBank mRNA AK074763.1 - 7 887
AK097765.1 - 1 416
AK309114.1 - 5 862
BC001327.1 - 7 897 3456988
BC001676.2 - 7 882 2963821
BC007265.1 - 7 905 3138466
BC007437.2 - 7 930 2961581
BT019999.1 - 7 749
U09585.1 - 7 747
Y12395.1 - 10 1247

View Full Product Details