Gene Transcripts

Gene Symbol
Nat9
Entrez Gene ID
303669
Gene Name
N-acetyltransferase 9 (GCN5-related, putative)
Gene Aliases
RGD1307096
Chromosome Location
Chr. 10: 103730215 - 103737050 on Build Rnor_6.0
UniGene ID
Rn.19920
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001134835.1 NP_001128307.1 5 886
XM_006247714.2 XP_006247776.1 6 745
GenBank mRNA BC158710.1 - 5 886

View Full Product Details