Gene Transcripts

Gene Symbol
Cd7
Entrez Gene ID
303747
Gene Name
Cd7 molecule
Gene Aliases
Chromosome Location
Chr. 10: 110229931 - 110232812 on Build Rnor_6.0
UniGene ID
Rn.8175
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001107074.1 NP_001100544.1 2 216
GenBank mRNA BC167030.1 - 2 188
FQ226313.1 - 2 218
FQ226643.1 - 2 218
FQ227263.1 - 2 218

View Full Product Details