Gene Transcripts

Gene Symbol
Mpa2l
Entrez Gene ID
305139
Gene Name
macrophage activation 2 like
Gene Aliases
RGD1561523
Chromosome Location
Chr. 14: 5927579 - 5946247 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_006250609.2 XP_006250671.2 9 2773

View Full Product Details