Gene Transcripts

Gene Symbol
Guf1
Entrez Gene ID
305317
Gene Name
GUF1 GTPase homolog (S. cerevisiae)
Gene Aliases
RGD1310774
Chromosome Location
Chr. 14: 62609484 - 62628862 on Build Rnor_6.0
UniGene ID
Rn.12097
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001107215.1 NP_001100685.1 6, 7 781
XM_008770207.1 XP_008768429.1 6, 7 1184

View Full Product Details