Gene Transcripts

Gene Symbol
Ctsm
Entrez Gene ID
306720
Gene Name
cathepsin M
Gene Aliases
Chromosome Location
Chr. 17: 3820606 - 3826015 on Build Rnor_6.0
UniGene ID
Rn.23458
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_181378.2 NP_852043.1 6 831
GenBank mRNA AF456462.1 - 6 831

View Full Product Details