Gene Transcripts

Gene Symbol
Frem3
Entrez Gene ID
307767
Gene Name
FRAS1 related extracellular matrix 3
Gene Aliases
RGD1562701
Chromosome Location
Chr. 19: 30973329 - 31031817 on Build Rnor_6.0
UniGene ID
Rn.41880
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001191699.1 NP_001178628.1 1 1585

View Full Product Details