Gene Transcripts

Gene Symbol
FZD8
Entrez Gene ID
8325
Gene Name
frizzled class receptor 8
Gene Aliases
FZ-8, hFZ8
Chromosome Location
Chr. 10: 35638249 - 35641434 on Build GRCh38
UniGene ID
Hs.302634
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_031866.2 NP_114072.1 1 450
GenBank mRNA AB043703.1 - 1 450

View Full Product Details