Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001107464.2 | NP_001100934.2 | 4 | 2007 | |
XM_006227960.2 | XP_006228022.1 | 4 | 1938 | ||
XM_006227962.2 | XP_006228024.1 | 4 | 2457 | ||
XM_006227964.2 | XP_006228026.1 | 4 | 2062 | ||
XM_006227965.2 | XP_006228027.1 | 4 | 2448 | ||
XM_008758760.1 | XP_008756982.1 | 4 | 2486 | ||
GenBank mRNA | FQ215550.1 | - | 1 | 676 |