Gene Transcripts
- Gene Symbol
- Cic
- Entrez Gene ID
- 308435
- Gene Name
- capicua transcriptional repressor
- Gene Aliases
- Chromosome Location
- Chr. 1: 82135428 - 82163007 on Build Rnor_6.0
- UniGene ID
- Rn.15668
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001107490.3 |
|
18, 19 |
4321 |
|
XM_008758933.1 |
XP_008757155.1
|
19, 20 |
7838 |
|
XM_008758934.1 |
XP_008757156.1
|
19, 20 |
8102 |
|
XM_008758935.1 |
XP_008757157.1
|
19, 20 |
7835 |
|
XM_008758936.1 |
XP_008757158.1
|
19, 20 |
7835 |
|
XM_008758937.1 |
XP_008757159.1
|
19, 20 |
7835 |
|
XM_008758938.1 |
XP_008757160.1
|
18, 19 |
4520 |
|
XM_008758939.1 |
XP_008757161.1
|
18, 19 |
4515 |
|
XM_008758940.1 |
XP_008757162.1
|
18, 19 |
4512 |
|
GenBank mRNA |
CK600637.1 |
- |
2, 3 |
280 |
|
View Full Product Details