Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_003561.2 | NP_003552.1 | 4 | 773 | |
NR_133651.1 | 5 | 889 | |||
GenBank mRNA | BC069539.1 | - | 4 | 613 | |
BC106731.1 | - | 4 | 773 | 40015660 | |
BC106732.1 | - | 4 | 773 | 40015664 | |
BC111804.1 | - | 4 | 773 | 40069347 | |
CR456885.1 | - | 4 | 429 | ||
U95301.1 | - | 4 | 869 |