Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_003986.2 | NP_003977.1 | 3 | 572 | |
XM_005253159.2 | XP_005253216.1 | 3 | 820 | ||
XM_005253160.2 | XP_005253217.1 | 3 | 817 | ||
XM_005253161.2 | XP_005253218.1 | 3 | 569 | ||
XM_011520402.1 | XP_011518704.1 | 4 | 980 | ||
XM_017018403.1 | XP_016873892.1 | 2 | 502 | ||
GenBank mRNA | AF082868.1 | - | 2 | 270 | |
AK290108.1 | - | 2 | 317 | ||
AK313422.1 | - | 2 | 320 | ||
BC011034.1 | - | 1 | 260 | 4042355 | |
CR456812.1 | - | 1 | 204 |