Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001008350.1 | NP_001008351.1 | 2 | 301 | |
XM_006256191.2 | XP_006256253.1 | 3 | 1131 | ||
XM_006256192.2 | XP_006256254.1 | 3 | 932 | ||
XM_008772812.1 | XP_008771034.1 | 3 | 915 | ||
GenBank mRNA | BC086421.1 | - | 2 | 301 |