Gene Transcripts

Gene Symbol
Tasp1
Entrez Gene ID
311468
Gene Name
taspase, threonine aspartase 1
Gene Aliases
RGD1308591
Chromosome Location
Chr. 3: 132888785 - 133131189 on Build Rnor_6.0
UniGene ID
Rn.22314
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001044243.1 NP_001037708.1 1 129
XM_008762262.1 XP_008760484.1 2 181
GenBank mRNA BC105618.1 - 1 129

View Full Product Details