Gene Transcripts

Gene Symbol
PRSS12
Entrez Gene ID
8492
Gene Name
protease, serine 12
Gene Aliases
BSSP-3, BSSP3, MRT1
Chromosome Location
Chr. 4: 118280038 - 118353042 on Build GRCh38
UniGene ID
Hs.445857
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_003619.3 NP_003610.2 7 1486
XM_005263318.4 XP_005263375.1 7 1759
XM_011532387.2 XP_011530689.1 7 1761
GenBank mRNA AJ001531.1 - 7 1479
BC007761.2 - 7 1486 4126901

View Full Product Details