Gene Transcripts

Gene Symbol
Rprd1b
Entrez Gene ID
311591
Gene Name
regulation of nuclear pre-mRNA domain containing 1B
Gene Aliases
Crept, RGD1304782
Chromosome Location
Chr. 3: 154507029 - 154553312 on Build Rnor_6.0
UniGene ID
Rn.4144
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001098727.1 NP_001092197.1 6, 7 1198
XM_006235412.1 XP_006235474.1 6, 7 1201
GenBank mRNA BC168699.1 - 6, 7 1203
CK365545.1 - 4, 5 446

View Full Product Details