Gene Transcripts

Gene Symbol
Murc
Entrez Gene ID
313225
Gene Name
muscle-related coiled-coil protein
Gene Aliases
RGD1310395
Chromosome Location
Chr. 5: 64413005 - 64420849 on Build Rnor_6.0
UniGene ID
Rn.218700
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001107931.2 NP_001101401.1 2 637
GenBank mRNA EU487255.1 - 2 637

View Full Product Details