Gene Transcripts

Gene Symbol
Tbc1d2
Entrez Gene ID
313234
Gene Name
TBC1 domain family, member 2
Gene Aliases
Chromosome Location
Chr. 5: 62213001 - 62261468 on Build Rnor_6.0
UniGene ID
Rn.168194
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001107933.1 NP_001101403.1 1 198
XM_006238056.2 XP_006238118.1 1 1577
GenBank mRNA BC168982.1 - 1 182

View Full Product Details