Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001316361.2 | NP_001303290.1 | 5 | 1178 | |
NM_001323347.1 | NP_001310276.1 | 5 | 891 | ||
NM_003657.3 | NP_003648.2 | 5 | 1178 | ||
XM_005260587.2 | XP_005260644.1 | 5 | 958 | ||
XM_005260589.2 | XP_005260646.1 | 5 | 958 | ||
XM_005260590.2 | XP_005260647.1 | 5 | 958 | ||
XM_005260591.2 | XP_005260648.1 | 5 | 958 | ||
XM_005260593.2 | XP_005260650.1 | 5 | 958 | ||
XM_005260594.2 | XP_005260651.1 | 5 | 958 | ||
XM_005260595.2 | XP_005260652.1 | 5 | 958 | ||
XM_011529090.2 | XP_011527392.1 | 5 | 886 | ||
XM_011529091.2 | XP_011527393.1 | 5 | 958 | ||
XM_017028110.1 | XP_016883599.1 | 5 | 958 | ||
XM_017028111.1 | XP_016883600.1 | 5 | 958 | ||
XM_017028112.1 | XP_016883601.1 | 5 | 958 | ||
GenBank mRNA | AF041260.1 | - | 5 | 957 | |
AK225656.1 | - | 5 | 947 | ||
AK294150.1 | - | 1 | 264 | ||
AK295318.1 | - | 2 | 357 | ||
AK308384.1 | - | 5 | 946 | ||
AK311215.1 | - | 2 | 360 | ||
AK315220.1 | - | 5 | 1178 | ||
AK316038.1 | - | 2 | 250 | ||
BC024044.1 | - | 5 | 959 | 4645428 | |
BC126346.1 | - | 5 | 889 | 8992062 | |
BC126348.1 | - | 5 | 889 | 8992064 | |
CR749643.1 | - | 1 | 20 |