Gene Transcripts

Gene Symbol
C8b
Entrez Gene ID
313421
Gene Name
complement component 8, beta polypeptide
Gene Aliases
Chromosome Location
Chr. 5: 124298271 - 124338055 on Build Rnor_6.0
UniGene ID
Rn.110603
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001191759.1 NP_001178688.1 4, 5 538
XM_006238495.2 XP_006238557.1 5, 6 858
GenBank mRNA BC090023.1 - 1, 2 63

View Full Product Details