Gene Transcripts

Gene Symbol
Ago1
Entrez Gene ID
313594
Gene Name
argonaute RISC catalytic component 1
Gene Aliases
Eif2c1
Chromosome Location
Chr. 5: 144444390 - 144479503 on Build Rnor_6.0
UniGene ID
Rn.62413
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_008764025.1 XP_008762247.1 18 2628
GenBank mRNA FQ234926.1 - 11 1397

View Full Product Details