Gene Transcripts
- Gene Symbol
- Grhl1
- Entrez Gene ID
- 313993
- Gene Name
- grainyhead-like 1 (Drosophila)
- Gene Aliases
- Dnmt3a
- Chromosome Location
- Chr. 6: 43734870 - 43795757 on Build Rnor_6.0
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
XM_234006.8 |
XP_234006.5
|
4 |
464 |
|
View Full Product Details