Gene Transcripts

Gene Symbol
PDLIM4
Entrez Gene ID
8572
Gene Name
PDZ and LIM domain 4
Gene Aliases
RIL
Chromosome Location
Chr. 5: 132257658 - 132273454 on Build GRCh38
UniGene ID
Hs.424312
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001131027.1 NP_001124499.1 6 1688
NM_003687.3 NP_003678.2 7 1806
XM_017010002.1 XP_016865491.1 5 1559
GenBank mRNA AF153882.1 - 7 1767

View Full Product Details