Gene Transcripts

Gene Symbol
Ccdc67
Entrez Gene ID
315438
Gene Name
coiled-coil domain containing 67
Gene Aliases
RGD1306632
Chromosome Location
Chr. 8: 14094742 - 14157235 on Build Rnor_6.0
UniGene ID
Rn.83901
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001014087.1 NP_001014109.2 10 1222
XM_008765933.1 XP_008764155.1 10 1405
XM_008765934.1 XP_008764156.1 10 1338
XM_008765935.1 XP_008764157.1 9 1447
XM_008765936.1 XP_008764158.1 10 1405
XM_008765937.1 XP_008764159.1 10 1403
XM_008765938.1 XP_008764160.1 10 1403
GenBank mRNA BC085333.1 - 10 1222

View Full Product Details