Gene Transcripts

Gene Symbol
Cd3e
Entrez Gene ID
315609
Gene Name
CD3 molecule, epsilon
Gene Aliases
Chromosome Location
Chr. 8: 49297604 - 49309369 on Build Rnor_6.0
UniGene ID
Rn.96221
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_008766161.1 XP_008764383.1 754
GenBank mRNA FQ228137.1 - 174

View Full Product Details