Gene Transcripts

Gene Symbol
Tbcc
Entrez Gene ID
316221
Gene Name
tubulin folding cofactor C
Gene Aliases
Chromosome Location
Chr. 9: 16405187 - 16406338 on Build Rnor_6.0
UniGene ID
Rn.18774
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001108200.2 NP_001101670.1 1 913
GenBank mRNA BC166467.1 - 1 913

View Full Product Details