Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001037808.2 | NP_001032897.1 | 19 | 2654 | |
NM_001199142.1 | NP_001186071.1 | 19 | 2774 | ||
NM_001267574.2 | NP_001254503.1 | 18 | 2743 | ||
NM_001286478.1 | NP_001273407.1 | 19 | 2575 | ||
NM_003752.4 | NP_003743.1 | 19 | 2605 | ||
XM_017023814.1 | XP_016879303.1 | 19 | 2560 | ||
GenBank mRNA | AK000739.1 | - | 18 | 2635 | |
AK055366.1 | - | 19 | 2474 | ||
AK130531.1 | - | 12 | 1802 | ||
AK292155.1 | - | 19 | 2774 | ||
AK293264.1 | - | 17 | 2183 | ||
AK299421.1 | - | 15 | 2014 | ||
AK301183.1 | - | 18 | 2513 | ||
AK301233.1 | - | 16 | 2101 | ||
AK303790.1 | - | 19 | 2518 | ||
AK304500.1 | - | 18 | 2284 | ||
AY762099.1 | - | 19 | 2517 | ||
BC000533.2 | - | 9 | 1106 | ||
BC001571.1 | - | 19 | 2539 | 3452953 | |
BC071705.1 | - | 19 | 2533 | 5552014 | |
BC157842.1 | - | 19 | 2663 | 9057061 | |
BC157849.1 | - | 18 | 2503 | 9057068 | |
BT007335.1 | - | 5 | 709 | ||
BX648075.1 | - | 11 | 3607 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001099661.2 | NP_001093131.1 | 19 | 2609 | |
NM_001317856.1 | NP_001304785.1 | 19 | 2654 | ||
NM_001317857.1 | NP_001304786.1 | 19 | 2605 | ||
XM_017023620.1 | XP_016879109.1 | 19 | 2561 | ||
XM_017023621.1 | XP_016879110.1 | 12 | 1736 |