Gene Transcripts

Gene Symbol
Clptm1l
Entrez Gene ID
316916
Gene Name
CLPTM1-like
Gene Aliases
RGD1307896
Chromosome Location
Chr. 1: 32281207 - 32297195 on Build Rnor_6.0
UniGene ID
Rn.101957
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001108240.1 NP_001101710.1 12 1469
XM_006227791.2 XP_006227853.1 12 1471

View Full Product Details