Gene Transcripts

Gene Symbol
KRT37
Entrez Gene ID
8688
Gene Name
keratin 37
Gene Aliases
HA7, K37, KRTHA7
Chromosome Location
Chr. 17: 41420557 - 41424570 on Build GRCh38
UniGene ID
Hs.673852
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_003770.4 NP_003761.3 7 1496
GenBank mRNA AJ786655.1 - 7 1332

View Full Product Details