Gene Transcripts
- Gene Symbol
- Ptprq
- Entrez Gene ID
- 360417
- Gene Name
- protein tyrosine phosphatase, receptor type, Q
- Gene Aliases
- rPTP-GMC1
- Chromosome Location
- Chr. 7: 49763657 - 50034932 on Build Rnor_6.0
- UniGene ID
- Rn.30011
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_022925.1 |
NP_075214.1
|
19 |
3068 |
|
GenBank mRNA |
AF063249.1 |
- |
19 |
3068 |
|
View Full Product Details