Gene Transcripts

Gene Symbol
Traf7
Entrez Gene ID
360491
Gene Name
TNF receptor-associated factor 7, E3 ubiquitin protein ligase
Gene Aliases
RGD1559653
Chromosome Location
Chr. 10: 13874456 - 13893091 on Build Rnor_6.0
UniGene ID
Rn.163128
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001127548.1 NP_001121020.1 18 1796
XM_006245982.2 XP_006246044.1 18 1890
XM_006245983.2 XP_006246045.1 18 1885
XM_006245984.2 XP_006246046.1 17 1731
XM_006245985.2 XP_006246047.1 17 1708
XM_006245986.2 XP_006246048.1 17 1728
XM_006245987.2 XP_006246049.1 17 1705
XM_006245988.2 XP_006246050.1 18 1927
XM_006245989.2 XP_006246051.1 18 1924
XM_006245990.2 XP_006246052.1 17 1677
XM_006245991.2 XP_006246053.1 17 1681
XM_006245992.2 XP_006246054.1 17 1674
XM_006245993.2 XP_006246055.1 16 1667
XM_008767584.1 XP_008765806.1 18 1943
XM_008767585.1 XP_008765807.1 18 1940
XM_008767586.1 XP_008765808.1 18 1938
GenBank mRNA BC161916.1 - 18 1796

View Full Product Details