Gene Transcripts

Gene Symbol
Rnmtl1
Entrez Gene ID
360569
Gene Name
RNA methyltransferase like 1
Gene Aliases
RGD1309077
Chromosome Location
Chr. 10: 64398339 - 64405017 on Build Rnor_6.0
UniGene ID
Rn.83045
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001108282.1 NP_001101752.1 2 360
GenBank mRNA FQ232324.1 - 2 358

View Full Product Details