Gene Transcripts

Gene Symbol
Med13l
Entrez Gene ID
360817
Gene Name
mediator complex subunit 13-like
Gene Aliases
Thrap2
Chromosome Location
Chr. 12: 43421317 - 43576863 on Build Rnor_6.0
UniGene ID
Rn.41407
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_006249420.2 XP_006249482.1 4 386
XM_006249421.2 XP_006249483.1 5 760
XM_006249422.2 XP_006249484.1 5 760
GenBank mRNA GU566026.1 - 5 745

View Full Product Details