Gene Transcripts
- Gene Symbol
- Fbxo21
- Entrez Gene ID
- 360818
- Gene Name
- F-box protein 21
- Gene Aliases
- Chromosome Location
- Chr. 12: 44244457 - 44279044 on Build Rnor_6.0
- UniGene ID
- Rn.137498
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
CB615901.1 |
- |
2 |
203 |
|
FQ212189.1 |
- |
11 |
1601 |
|
View Full Product Details