Gene Transcripts

Gene Symbol
Kif14
Entrez Gene ID
360849
Gene Name
kinesin family member 14
Gene Aliases
Chromosome Location
Chr. 13: 53350073 - 53421992 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_008769585.1 XP_008767807.1 2 789

View Full Product Details