Gene Transcripts
- Gene Symbol
- Tbc1d14
- Entrez Gene ID
- 360956
- Gene Name
- TBC1 domain family, member 14
- Gene Aliases
- SRF-2, UR-NR#2
- Chromosome Location
- Chr. 14: 79332054 - 79438765 on Build Rnor_6.0
- UniGene ID
- Rn.203187
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001012152.2 |
NP_001012152.2
|
7, 8 |
739 |
|
NM_001034021.2 |
NP_001029193.2
|
9, 10 |
1601 |
|
NM_001113365.1 |
NP_001106836.1
|
10, 11 |
1828 |
|
XM_006251133.2 |
XP_006251195.1
|
10, 11 |
1834 |
|
XM_006251134.2 |
XP_006251196.1
|
9, 10 |
1526 |
|
XM_008770323.1 |
XP_008768545.1
|
10, 11 |
2015 |
|
XM_008770324.1 |
XP_008768546.1
|
8, 9 |
3523 |
|
XM_008770325.1 |
XP_008768547.1
|
7, 8 |
676 |
|
XM_008770326.1 |
XP_008768548.1
|
7, 8 |
719 |
|
GenBank mRNA |
AB108668.1 |
- |
7, 8 |
676 |
|
AB117752.1 |
- |
10, 11 |
1828 |
|
BC099760.1 |
- |
9, 10 |
1574 |
7930485 |
View Full Product Details