Gene Transcripts

Gene Symbol
Arhgef40
Entrez Gene ID
361034
Gene Name
Rho guanine nucleotide exchange factor (GEF) 40
Gene Aliases
RGD1308093
Chromosome Location
Chr. 15: 28377934 - 28403090 on Build Rnor_6.0
UniGene ID
Rn.98243
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001271313.1 NP_001258242.1 6, 7 1879
XM_008770553.1 XP_008768775.1 6, 7 1941
XM_008770554.1 XP_008768776.1 6, 7 1941

View Full Product Details