Gene Transcripts

Gene Symbol
Commd10
Entrez Gene ID
361323
Gene Name
COMM domain containing 10
Gene Aliases
Chromosome Location
Chr. 18: 41022491 - 41154971 on Build Rnor_6.0
UniGene ID
Rn.163373
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001004276.1 NP_001004276.1 6 572
XM_006254697.2 XP_006254759.1 5 525
GenBank mRNA BC079373.1 - 6 572

View Full Product Details